Anti-P53 Proteinwt-p53 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-0033R-A350
BOSSBS-0033R-A350EA 901 EUR
BOSSBS-0033R-A350
Anti-P53 Proteinwt-p53 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Acts as a tumor suppressor in many tumor types; induces growth arrest or apoptosis depending on the physiological circumstances and cell type. Involved in cell cycle regulation as a trans-activator that acts to negatively regulate cell division by controlling a set of genes required for this process. One of the activated genes is an inhibitor of cyclin-dependent kinases. Apoptosis induction seems to be mediated either by stimulation of BAX and FAS antigen expression, or by repression of Bcl-2 expression. In cooperation with mitochondrial PPIF is involved in activating oxidative stress-More Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol P53 Proteinwt-p53
Environmentally Preferable
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 350
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 7157
Isotyp IgG
ImmunoChemie Yes
ImmunFluoreszenz Yes
Immunogen 240-290/393
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR