Anti-MLANA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-0051R-A350
BOSSBS-0051R-A350EA 893 EUR
BOSSBS-0051R-A350
Anti-MLANA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MLANA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR