Anti-INS Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-0056R-A647
BOSSBS-0056R-A647EA 876 EUR
BOSSBS-0056R-A647
Anti-INS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: INS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR