Anti-APAF1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0059R-CY5.5
BOSSBS-0059R-CY5.5EA 900 EUR
BOSSBS-0059R-CY5.5
Anti-APAF1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Oligomeric Apaf-1 mediates the cytochrome c-dependent autocatalytic activation of pro-caspase-9 (Apaf-3), leading to the activation of caspase-3 and apoptosis. This activation requires ATP. Isoform 6 is less effective in inducing apoptosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APAF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat