Anti-MUC16 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-0091R-A350
BOSSBS-0091R-A350EA 901 EUR
BOSSBS-0091R-A350
Anti-MUC16 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MUC16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human