Anti-PR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0111R-A750
BOSSBS-0111R-A750EA 899 EUR
BOSSBS-0111R-A750
Anti-PR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The steroid hormones and their receptors are involved in the regulation of eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues. Progesterone receptor isoform B (PRB) is involved activation of c-SRC/MAPK signaling on hormone stimulation. Isoform A: inactive in stimulating c-Src/MAPK signaling on hormone stimulation. Isoform 4: Increases mitochondrial membrane potential and cellular respiration upon stimulation by progesterone.

Type: Primary
Antigen: Progesterone Receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/933
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR