Anti-SARS ORF1a polyProtein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-0132R-A680
BOSSBS-0132R-A680EA 1130 EUR
BOSSBS-0132R-A680
Anti-SARS ORF1a polyProtein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Severe acute respiratory syndrome (SARS) is a viral respiratory illness caused by a coronavirus, called SARS-associated coronavirus (SARS-CoV). Human coronaviruses (HCoVs) were previously only associated with mild diseases. The SARS-CoV genome contains five major open reading frames (ORFs) that encode the replicase polyprotein; the spike (S), envelope (E), and membrane (M) glycoproteins; and the nucleocapsid protein (N).
Type: Primary
Antigen: SARS ORF1a polyprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 3501-3600/7073
Host: Rabbit
Isotype: IgG
Reactivity: Virus