Anti-SARS ORF1a polyProtein Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0132R-A750
BOSSBS-0132R-A750EA 1130 EUR
BOSSBS-0132R-A750
Anti-SARS ORF1a polyProtein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Severe acute respiratory syndrome (SARS) is a viral respiratory illness caused by a coronavirus, called SARS-associated coronavirus (SARS-CoV). Human coronaviruses (HCoVs) were previously only associated with mild diseases. The SARS-CoV genome contains five major open reading frames (ORFs) that encode the replicase polyprotein; the spike (S), envelope (E), and membrane (M) glycoproteins; and the nucleocapsid protein (N).

Type: Primary
Antigen: SARS ORF1a polyprotein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 3501-3600/7073
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR