Anti-GCH1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-0136R-CY5
BOSSBS-0136R-CY5EA 900 EUR
BOSSBS-0136R-CY5
Anti-GCH1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GTP cyclohydrolase I (also designated dopa-responsive dystonia) catalyzes the conversion of GTP to D-erythro-7,8-dihydroneopterin triphosphate, the first and rate-limiting step in tetrahydrobiopterin (BH4) biosynthesis. Tetrahydrobiopterin is an essential cofactor for 3 aromatic amino acid monooxygenases: phenylalanine, tyrosine, and tryptophan hydroxylases. Animals can synthesize tetrahydrobiopterin in vivo from GTP through several enzymatic reactions.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GTP cyclohydrolase 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat