Anti-DDC Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-0180R-A488
BOSSBS-0180R-A488EA 901 EUR
BOSSBS-0180R-A488
Anti-DDC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Catalyzes the decarboxylation of L-3,4-dihydroxyphenylalanine (DOPA) to dopamine, L-5-hydroxytryptophan to serotonin and L-tryptophan to tryptamine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DDC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR