Anti-DOPA Decarboxylase Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0180R-A680
BOSSBS-0180R-A680EA 899 EUR
BOSSBS-0180R-A680
Anti-DOPA Decarboxylase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
catalyses the decarboxylation of L-3,4-dihydroxyphenylalanine (DOPA) to dopamine, L-5-hydroxytryptophan to serotonin and L-tryptophan to tryptamine.

Type: Primary
Antigen: DOPA Decarboxylase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/480
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR