Anti-Nociceptin Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0181R-A680
BOSSBS-0181R-A680EA 890 EUR
BOSSBS-0181R-A680
Anti-Nociceptin Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
G-protein coupled opioid receptor that functions as receptor for the endogenous neuropeptide nociceptin. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. Signaling via G proteins mediates inhibition of adenylate cyclase activity and calcium channel activity. Arrestins modulate signaling via G proteins and mediate the activation of alternative signaling pathways that lead to the activation of MAP kinases. Plays a role in modulating nociception and the perception of pain. Plays a role in the regulation of locomotor activity by the neuropeptide nociceptin.

Type: Primary
Antigen: Nociceptin receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-47/370
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR