Anti-VIPR2 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-0197R
BOSSBS-0197REA 752 EUR
BOSSBS-0197R
Anti-VIPR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VIPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR