Anti-ACEI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-0203R-HRP
BOSSBS-0203R-HRPEA 900 EUR
BOSSBS-0203R-HRP
Anti-ACEI Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Angiotensin I converting Enzyme Inhibitor, ACEI also designated Bradykinin-potentiating peptide. The peptide: PTHIKWGD. Processed active peptide.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ACEI
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR