Anti-PPAR delta Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0250R-A680
BOSSBS-0250R-A680EA 890 EUR
BOSSBS-0250R-A680
Anti-PPAR delta Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Ligand-activated transcription factor. Receptor that binds peroxisome proliferators such as hypolipidemic drugs and fatty acids. Has a preference for poly-unsaturated fatty acids, such as gamma-linoleic acid and eicosapentanoic acid. Once activated by a ligand, the receptor binds to promoter elements of target genes. Regulates the peroxisomal beta-oxidation pathway of fatty acids. Functions as transcription activator for the acyl-CoA oxidase gene. Decreases expression of NPC1L1 once activated by a ligand (By similarity).

Type: Primary
Antigen: PPAR delta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 2-100/440
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR