Anti-PBEF CT Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-0273R-A555
BOSSBS-0273R-A555EA 893 EUR
BOSSBS-0273R-A555
Anti-PBEF CT Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
icotinamide phosphoribosyltransferase (NAmPRTase) (Nampt) (Pre-B cell-enhancing factor) (Pre-B-cell colony-enhancing factor 1) (Fisfatin) Catalyzes the condensation of nicotinamide with5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. Expressed in large amounts in bone marrow, liver tissue, and muscle. Also present in heart, placenta, lung, and kidney tissues. Subcellular Location is in Cytoplasm. Belongs to the NAPRTase family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PBEF CT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR