Anti-PBEF CT Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-0273R-CY7
BOSSBS-0273R-CY7EA 900 EUR
BOSSBS-0273R-CY7
Anti-PBEF CT Rabbit Polyclonal Antibody (Cy7®)
Antikörper
icotinamide phosphoribosyltransferase (NAmPRTase) (Nampt) (Pre-B cell-enhancing factor) (Pre-B-cell colony-enhancing factor 1) (Fisfatin) Catalyzes the condensation of nicotinamide with5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. Expressed in large amounts in bone marrow, liver tissue, and muscle. Also present in heart, placenta, lung, and kidney tissues. Subcellular Location is in Cytoplasm. Belongs to the NAPRTase family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PBEF CT
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat