Anti-IGBP1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-0298R-CY3
BOSSBS-0298R-CY3EA 909 EUR
BOSSBS-0298R-CY3
Anti-IGBP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Associated to surface IgM-receptor; may be involved in signal transduction. Involved in regulation of the catalytic activity of the phosphatases PP2A, PP4 and PP6 by protecting their partially folded catalytic subunits from degradative polyubiquitination until they associate with regulatory subunits.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IGBP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat