Anti-PADI4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0318R-A750
BOSSBS-0318R-A750EA 890 EUR
BOSSBS-0318R-A750
Anti-PADI4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PADI4 (peptidyl arginine deiminase, type IV) catalyses the deimination of arginine residues of proteins. Down-regulates histone H3 and H4 arginine methylation, both by preventing arginine methylation by CARM1 and HRMT1L2/PRMT1 and by converting methylarginine to citrulline. subcellular location at cytoplasmic granules. Belongs to the protein arginine deiminase family. This gene may play a role in granulocyte and macrophage development leading to inflammation and immune response.

Type: Primary
Antigen: PADI4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-500/663
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR