Anti-factor 8/F8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0332R-A750
BOSSBS-0332R-A750EA 899 EUR
BOSSBS-0332R-A750
Anti-factor 8/F8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Factor VIII, along with calcium and phospholipid, acts as a cofactor for factor IXa when it converts factor X to the activated form, factor Xa.

Type: Primary
Antigen: factor 8/F8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1451-1550/2351
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR