Anti-Protein A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-0362R-FITC
BOSSBS-0362R-FITCEA 900 EUR
BOSSBS-0362R-FITC
Anti-Protein A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Protein A is a 40-60 kDa surface protein originally found in the cell wall of the bacteria Staphylococcus aureus (SPA). SPA binds proteins from many of mammalian species, most notably IgGs, and helps inhibit phagocytic engulfment and acts as an immunological disguise via this type of interaction, thus the bacteria will disrupts opsonization and phagocytosis. SPA is known to bind with Fc region of immunoglobulins preferentially through interaction with the VH3 variable region of the heavy chain. SPA has been shown to bind with high affinity to human IgG1 and IgG2 as well as mouse IgG2a and IgG2b, whereas bind with moderate affinity to human IgM, IgA and IgE as well as mouse IgG3 and IgG1. SPA is often produced in E. coli and is practically coupled to other molecules such as enzymes, biotin, radioactive iodine for use in immunology and other biological research. SPA is also immobilized onto solid supports such as agarose beads for total IgG purifying or interest protein or protein complex identifying in immunoprecipitation studies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR