Anti-NT5C2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-0372R-A647
BOSSBS-0372R-A647EA 901 EUR
BOSSBS-0372R-A647
Anti-NT5C2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
May have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases. Preferentially hydrolyzes inosine 5'-monophosphate (IMP) and other purine nucleotides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NT5C2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR