Anti-TP-5/Thymopentin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-0392R-A680
BOSSBS-0392R-A680EA 890 EUR
BOSSBS-0392R-A680
Anti-TP-5/Thymopentin Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Sequence: H-Arg-Lys-Asp-Val-Tyr-OH
MW: 679,8.
Type: Primary
Antigen: TP-5/Thymopentin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen