Anti-TP5 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0392R-CY5.5
BOSSBS-0392R-CY5.5EA 900 EUR
BOSSBS-0392R-CY5.5
Anti-TP5 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Sequence:: H-Arg-Lys-Asp-Val-Tyr-OH
MW: 679.13
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TP5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat