Anti-TP5 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-0392R-CY7
BOSSBS-0392R-CY7EA 900 EUR
BOSSBS-0392R-CY7
Anti-TP5 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Sequence:: H-Arg-Lys-Asp-Val-Tyr-OH
MW: 679.14

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TP5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR