Anti-MMP9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-0397R-A647
BOSSBS-0397R-A647EA 901 EUR
BOSSBS-0397R-A647
Anti-MMP9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
May play an essential role in local proteolysis of the extracellular matrix and in leukocyte migration. Could play a role in bone osteoclastic resorption. Cleaves KiSS1 at a Gly-|-Leu bond. Cleaves type IV and type V collagen into large C-terminal three quarter fragments and shorter N-terminal one quarter fragments. Degrades fibronectin but not laminin or Pz-peptide.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat