Anti-MMP9 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-0397R-CY3
BOSSBS-0397R-CY3EA 901 EUR
BOSSBS-0397R-CY3
Anti-MMP9 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
May play an essential role in local proteolysis of the extracellular matrix and in leukocyte migration. Could play a role in bone osteoclastic resorption. Cleaves KiSS1 at a Gly-|-Leu bond. Cleaves type IV and type V collagen into large C-terminal three quarter fragments and shorter N-terminal one quarter fragments. Degrades fibronectin but not laminin or Pz-peptide.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR