Anti-Deltex 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-0399R-A750
BOSSBS-0399R-A750EA 890 EUR
BOSSBS-0399R-A750
Anti-Deltex 1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Functions as a ubiquitin ligase protein in vivo, mediating ubiquitination and promoting degradation of MEKK1, suggesting that it may regulate the Notch pathway via some ubiquitin ligase activity (By similarity). Regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. Mainly acts as a positive regulator of Notch, but it also acts as a negative regulator, depending on the developmental and cell context. Mediates the antineural activity of Notch, possibly by inhibiting the transcriptional activation mediated by MATCH1. Involved in neurogenesis, lymphogenesis and myogenesis, and may also be involved in MZB (Marginal zone B) cell differentiation. Promotes B-cell development at the expense of T-cell development, suggesting that it can antagonize NOTCH1.
Type: Primary
Antigen: Deltex 1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 351-450/620
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Others