Anti-Metal ion Transporter Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0401R-A680
BOSSBS-0401R-A680EA 890 EUR
BOSSBS-0401R-A680
Anti-Metal ion Transporter Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
No data available.

Type: Primary
Antigen: Metal ion transporter
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/276
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR