Anti-RGS2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-0492R-CY3
BOSSBS-0492R-CY3EA 900 EUR
BOSSBS-0492R-CY3
Anti-RGS2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. May play a role in leukemogenesis. Plays a role in negative feedback control pathway for adenylyl cyclase signaling. Binds EIF2B5 and blocks its activity, thereby inhibiting the translation of mRNA into protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR