Anti-ETFA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0494R-A680
BOSSBS-0494R-A680EA 890 EUR
BOSSBS-0494R-A680
Anti-ETFA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: ETFA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 184-260/333
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Xenopus laevis
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR