Anti-ADRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-0498R-A555
BOSSBS-0498R-A555EA 901 EUR
BOSSBS-0498R-A555
Anti-ADRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Beta-adrenergic receptors mediate the catecholamine-induced activation of adenylate cyclase through the action of G proteins. This receptor binds epinephrine and norepinephrine with approximately equal affinity. Mediates Ras activation through G(s)-alpha- and cAMP-mediated signaling.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADRB1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR