Anti-ADRB1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0498R-CY5.5
BOSSBS-0498R-CY5.5EA 901 EUR
BOSSBS-0498R-CY5.5
Anti-ADRB1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Beta-adrenergic receptors mediate the catecholamine-induced activation of adenylate cyclase through the action of G proteins. This receptor binds epinephrine and norepinephrine with approximately equal affinity. Mediates Ras activation through G(s)-alpha- and cAMP-mediated signaling.
Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADRB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat