Anti-SLC9A1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0505R-CY5.5
BOSSBS-0505R-CY5.5EA 900 EUR
BOSSBS-0505R-CY5.5
Anti-SLC9A1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Involved in pH regulation to eliminate acids generated by active metabolism or to counter adverse environmental conditions. Major proton extruding system driven by the inward sodium ion chemical gradient. Plays an important role in signal transduction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC9A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat