Anti-RARB Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-0516R-A555
BOSSBS-0516R-A555EA 893 EUR
BOSSBS-0516R-A555
Anti-RARB Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Receptor for retinoic acid. Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence or presence of hormone ligand, acts mainly as an activator of gene expression due to weak binding to corepressors. In concert with RARG, required for skeletal growth, matrix homeostasis and growth plate function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RARB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR