Anti-Centromeric histone H3-like Protein-2 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-0621R-CY5.5
BOSSBS-0621R-CY5.5EA 893 EUR
BOSSBS-0621R-CY5.5
Anti-Centromeric histone H3-like Protein-2 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Centromeric histone H3-like protein-2

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Centromeric histone H3-like Protein-2
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR