Anti-Dmt1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-0653R
BOSSBS-0653REA 725 EUR
BOSSBS-0653R
Anti-Dmt1 Rabbit Polyclonal Antibody
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Dmt1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR