Anti-DMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-0653R-A350
BOSSBS-0653R-A350EA 893 EUR
BOSSBS-0653R-A350
Anti-DMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Dmt1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR