Anti-DMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-0653R-A488
BOSSBS-0653R-A488EA 893 EUR
BOSSBS-0653R-A488
Anti-DMT1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Important in metal transport, in particular iron. Can also transport manganese, cobalt, cadmium, nickel, vanadium and lead. Involved in apical iron uptake into duodenal enterocytes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Dmt1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant