Anti-SLC33A1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0699R-A750
BOSSBS-0699R-A750EA 890 EUR
BOSSBS-0699R-A750
Anti-SLC33A1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Probable acetyl-CoA transporter necessary for O-acetylation of gangliosides (PubMed:9096318). Negatively regulates BMP signaling (PubMed:25402622).

Type: Primary
Antigen: SLC33A1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:481-549/549
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR