Anti-Collagen 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0709R-A680
BOSSBS-0709R-A680EA 899 EUR
BOSSBS-0709R-A680
Anti-Collagen 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Type II collagen is specific for cartilaginous tissues. It is essential for the normal embryonic development of the skeleton, for linear growth and for the ability of cartilage to resist compressive forces.

Type: Primary
Antigen: Collagen 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1201-1300/1487
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit, Guinea Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR