Anti-KCNMA1 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-0729R
BOSSBS-0729REA 752 EUR
BOSSBS-0729R
Anti-KCNMA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KCNMA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat