Anti-FGF8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-0735R-A555
BOSSBS-0735R-A555EA 901 EUR
BOSSBS-0735R-A555
Anti-FGF8 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system (By similarity). Cooperates with Wnt-1 in mouse mammary tumor virus-induced murine mammary tumorigenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR