Anti-FGF8 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-0735R-CY3
BOSSBS-0735R-CY3EA 909 EUR
BOSSBS-0735R-CY3
Anti-FGF8 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system (By similarity). Cooperates with Wnt-1 in mouse mammary tumor virus-induced murine mammary tumorigenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FGF8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat