Anti-Lpin1 Protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-0759R-A750
BOSSBS-0759R-A750EA 890 EUR
BOSSBS-0759R-A750
Anti-Lpin1 Protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyses the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis. Acts also as nuclear transcriptional coactivator for PPARGC1A/PPARA regulatory pathway to modulate lipid metabolism gene expression. Is involved in adipocyte differentiation. Isoform 1 is recruited at the mitochondrion outer membrane and is involved in mitochondrial fission by converting phosphatidic acid to diacylglycerol.
Type: Primary
Antigen: Lpin1 protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/924
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat