Anti-LPIN1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-0760R-CY3
BOSSBS-0760R-CY3EA 900 EUR
BOSSBS-0760R-CY3
Anti-LPIN1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis. Acts also as nuclear transcriptional coactivator for PPARGC1A/PPARA regulatory pathway to modulate lipid metabolism gene expression. Is involved in adipocyte differentiation. Isoform 1 is recruited at the mitochondrion outer membrane and is involved in mitochondrial fission by converting phosphatidic acid to diacylglycerol.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LPIN1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat