Anti-Casein Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-0813R-CY7
BOSSBS-0813R-CY7EA 883 EUR
BOSSBS-0813R-CY7
Anti-Casein Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Casein is the name for a family of related phosphoproteins. These proteins are commonly found in mammalian milk, making up 80% of the proteins in cow milk and between 20% and 45% of the proteins in human milk. Casein has a wide variety of uses, from being a major component of cheese, to use as a food additive, to a binder for safety matches. As a food source, casein supplies amino acids; carbohydrates; and two inorganic elements, calcium and phosphorus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Casein
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR