Anti-GRIA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-0837R-A350
BOSSBS-0837R-A350EA 876 EUR
BOSSBS-0837R-A350
Anti-GRIA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Glutathione reductase (GR) is a member of pyridine nucleotide- disulfideoxidoreductases, which includes the closely related enzymes thioredoxin reductase, lipoamide dehydrogenase, trypanothione reductase and mercuric ion reductase. GR is a cytoplasmic flavoenzyme widely distributed in aerobic organisms. The dimeric protein is composed of two identical subunits, each containing 1 FAD and 1 redox-active disulfide/dithiol as components of the catalytic apparatus. It plays a role in maintaining glutathione (GSH) in its reduced form by catalyzing the reduction of glutathione disulfide (GSSG): GSSG + NADPH + H+?2GSH + NADP+. In mosteukaryotic cells, GR maintains the ratio of [GSH]/[GSSG], and participates in several vital functions such as the detoxification of reactive oxygen species as well as protein and DNA biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLUR/glutathione reductase
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR