Anti-AMACR Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0840R-CY5.5
BOSSBS-0840R-CY5.5EA 893 EUR
BOSSBS-0840R-CY5.5
Anti-AMACR Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
alpha-methylacyl-CoA racemase(AMACR/P504S) is Prostate-specific antigen (PSA) screening for prostate cancer is now widespread in the United States among men of all ages. However PSA has limited specificity because benign disease, including prostatic enlargement and inflammation, can increase PSA levels. Thus, a more specific prostate cancer markers is needed. One such potential marker is AMACR, an enzyme that is involved in peroxisomal beta-oxidation of dietary branched-chain fatty acids. Recent studies have shown that, compared with expression in normal or benign prostate epithelium, AMACRMore Product Information