Anti-GFP Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-0890R-CY3
BOSSBS-0890R-CY3EA 893 EUR
BOSSBS-0890R-CY3
Anti-GFP Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Green fluorescence protein (GFP) is a 27 kDa protein derived from the jellyfish Aequorea victoria, which emits green light (emission peak at a wavelenth of 509 nm) when excited by blue light (excitation peak at a wavelenth of 395 nm). Green Fluorescent Protein (GFP) has become an invaluable tool in cell biology research, since its intrinsic fluorescence can be visualized in living cells. GFP fluorescence is stable under fixation conditions and suitable for a variety of applications. GFP has been widely used as a reporter for gene expression, enabling researchers to visualize and localize GFP-tagged proteins within living cells without the need for chemical staining. Other applications of GFP include assessment of protein protein interactions through the yeast two hybrid system and measurement of distance between proteins through fluorescence energy transfer (FRET) protocols. GFP technnology has considerably contributed to a greater understanding of cellular physiology.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Fish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR